The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175358.4 | NP_780567.1 | 11 - 12 | 1314 | 116 |
GenBank mRNA | AK029137.1 | - | 11 - 12 | 1054 | 116 |
AK032922.1 | - | 11 - 12 | 1035 | 116 | |
AK077949.1 | - | 11 - 12 | 1063 | 116 | |
AK132304.1 | - | 11 - 12 | 1316 | 116 | |
AK160360.1 | - | 11 - 12 | 1054 | 116 |