The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009044.2 | NP_033070.2 | 2 - 3 | 432 | 143 |
GenBank mRNA | AK031627.1 | - | 2 - 3 | 231 | 143 |
BC139447.1 | - | 2 - 3 | 372 | 143 | |
BC139448.1 | - | 2 - 3 | 372 | 143 | |
BC139770.1 | - | 2 - 3 | 374 | 143 | |
X60271.1 | - | 2 - 3 | 431 | 143 |