The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164598.1 | NP_001158070.1 | 1 - 2 | 999 | 102 |
GenBank mRNA | BC048951.1 | - | 1 - 2 | 102 |