Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map4
Entrez Gene ID
17758
Gene Name
microtubule-associated protein 4
Gene Aliases
AA407148, MAP-4, Mtap-4, Mtap4
Chromosome Location
Chr.9: 109931774 - 110083954 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205330.1 NP_001192259.1 8 - 9 2099 67
NM_001205332.1 NP_001192261.1 8 - 9 2099 67
NM_008633.4 NP_032659.2 8 - 9 2099 67
GenBank mRNA BC044654.1 - 8 - 9 2009 67
BC050893.1 - 8 - 9 1974 67
BC055332.1 - 8 - 9 1980 67
BC055364.1 - 7 - 8 1939 67
M72414.1 - 8 - 9 2403 67

View Full Product Details