Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifb2
Entrez Gene ID
66557
Gene Name
BPI fold containing family B, member 2
Gene Aliases
2310034L21Rik, 2310069A01Rik, AV089125, Bpil1, LPLUNC2, RYSR
Chromosome Location
Chr.2: 153875045 - 153895270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025631.3 NP_079907.2 8 - 9 780 88
GenBank mRNA AK009504.1 - 8 - 9 755 88
AK009619.1 - 8 - 9 753 88
AK009754.1 - 4 - 5 356 88
AK009776.1 - 8 - 9 737 88
AK010215.1 - 8 - 9 751 87
AK028156.1 - 8 - 9 751 88
AK028158.1 - 8 - 9 754 88
AK075874.1 - 8 - 9 751 88
AK160516.1 - 8 - 9 751 88
BC116681.1 - 8 - 9 735 88
BC116727.1 - 8 - 9 88

View Full Product Details