The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039939.1 | NP_001035028.1 | 7 - 8 | 758 | 83 |
GenBank mRNA | AK122413.1 | - | 7 - 8 | 758 | 83 |