The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194357.1 | NP_919338.2 | 13 - 14 | 1392 | 64 |
GenBank mRNA | AK132627.1 | - | 8 - 9 | 744 | 64 |
BC048084.1 | - | 13 - 14 | 1392 | 64 |