Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scarf1
Entrez Gene ID
380713
Gene Name
scavenger receptor class F, member 1
Gene Aliases
AA986099, SREC, SREC-I, mKIAA0149
Chromosome Location
Chr.11: 75513495 - 75526580 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004157.2 NP_001004157.2 7 - 8 1287 64
GenBank mRNA AK133477.1 - 7 - 8 1270 64
AK220215.1 - 7 - 8 1271 64
BC057600.1 - 6 - 7 1194 64
BC080288.1 - 7 - 8 1287 64

View Full Product Details