The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183263.5 | NP_899086.2 | 3 - 4 | 763 | 71 |
GenBank mRNA | AK012200.1 | - | 3 - 4 | 738 | 71 |
AK014739.1 | - | 3 - 4 | 729 | 71 | |
AK147805.1 | - | 3 - 4 | 744 | 71 | |
BC038288.1 | - | 3 - 4 | 712 | 71 | |
BC090975.1 | - | 3 - 4 | 735 | 71 | |
BC115622.1 | - | 3 - 4 | 489 | 71 | |
BC115623.1 | - | 3 - 4 | 489 | 71 |