The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027773.3 | NP_082049.1 | 1 - 2 | 199 | 68 |
GenBank mRNA | AK077374.1 | - | 1 - 2 | 159 | 68 |
AK160183.1 | - | 1 - 2 | 160 | 68 | |
BC115519.1 | - | 1 - 2 | 198 | 68 | |
BC115520.1 | - | 1 - 2 | 199 | 68 |