The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024918.1 | NP_001020089.1 | 14 - 15 | 1893 | 69 |
NM_027689.3 | NP_081965.2 | 10 - 11 | 1225 | 69 | |
GenBank mRNA | AB086957.1 | - | 14 - 15 | 1747 | 69 |
AB089184.1 | - | 10 - 11 | 1226 | 69 | |
AK034131.1 | - | 14 - 15 | 1880 | 69 | |
AK132673.1 | - | 11 - 12 | 1288 | 69 | |
AY102010.2 | - | 14 - 15 | 1776 | 69 | |
AY342003.1 | - | 14 - 15 | 1758 | 69 | |
BC138719.1 | - | 10 - 11 | 1213 | 69 | |
BC138720.1 | - | 10 - 11 | 1213 | 69 |