Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fads6
Entrez Gene ID
328035
Gene Name
fatty acid desaturase domain family, member 6
Gene Aliases
BC044804, OTTMUSG00000021749
Chromosome Location
Chr.11: 115279622 - 115297822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178035.4 NP_828874.3 2 - 3 400 71
GenBank mRNA AB087503.1 - 2 - 3 368 71
BC044804.1 - 2 - 3 258 71
BC132041.1 - 2 - 3 351 71
BC132067.1 - 2 - 3 351 71

View Full Product Details