The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009950.1 | NP_001009950.1 | 8 - 9 | 1115 | 81 |
GenBank mRNA | BC089013.1 | - | 8 - 9 | 1115 | 81 |