The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029428.1 | NP_083704.1 | 6 - 7 | 1020 | 80 |
GenBank mRNA | BC137980.1 | - | 6 - 7 | 1007 | 80 |