Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcng4
Entrez Gene ID
66733
Gene Name
potassium voltage-gated channel, subfamily G, member 4
Gene Aliases
4921535I01Rik, AW049024, KV6.3, KV6.4
Chromosome Location
Chr.8: 119623854 - 119636353 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025734.2 NP_080010.2 1 - 2 446 63
GenBank mRNA AF450109.1 - 1 - 2 112 63
BC043936.1 - 1 - 2 446 63

View Full Product Details