The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153145.4 | NP_694785.3 | 8 - 9 | 1205 | 134 |
GenBank mRNA | AB255881.1 | - | 7 - 8 | 1124 | 134 |
AB255882.1 | - | 8 - 9 | 1212 | 134 | |
AF498360.1 | - | 7 - 8 | 1130 | 134 | |
AY732492.1 | - | 7 - 8 | 1130 | 134 |