The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013851.2 | NP_038879.2 | 37 - 38 | 4922 | 82 |
GenBank mRNA | AF213393.1 | - | 4 - 5 | 415 | 82 |
AF498362.1 | - | 37 - 38 | 4922 | 82 | |
AK043744.1 | - | - | 471 | 82 | |
AK173034.1 | - | 36 - 37 | 4663 | 82 | |
BC089557.1 | - | 13 - 14 | 1565 | 82 | |
BC141401.1 | - | 37 - 38 | 4885 | 82 |