Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nwd1
Entrez Gene ID
319555
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
A230063L24Rik
Chromosome Location
Chr.8: 72646225 - 72716880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176940.5 NP_795914.3 10 - 11 2914 65
GenBank mRNA AK034868.1 - - 2613 65
AK038792.1 - 9 - 10 2607 65
AK087622.1 - 10 - 11 2919 65
AK162888.1 - 10 - 11 2909 65
BC082552.1 - 10 - 11 2854 65
BC144786.1 - 8 - 9 2504 65
BC144787.1 - 8 - 9 2504 65
BC145703.1 - 8 - 9 2504 65

View Full Product Details