Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nwd1
Entrez Gene ID
319555
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
A230063L24Rik
Chromosome Location
Chr.8: 72646225 - 72716880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176940.5 NP_795914.3 2 - 3 387 82
GenBank mRNA AK034868.1 - - 384 82
AK038792.1 - 2 - 3 377 82
AK087622.1 - 2 - 3 388 82
AK162888.1 - 2 - 3 381 82
BC082552.1 - 2 - 3 327 82
BY731616.1 - 2 - 3 384 82

View Full Product Details