Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trp53i13
Entrez Gene ID
216964
Gene Name
transformation related protein 53 inducible protein 13
Gene Aliases
2410019G02Rik, Tp53i13
Chromosome Location
Chr.11: 77508099 - 77515921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024920.1 NP_001020091.1 2 - 3 186 80
GenBank mRNA AK213971.1 - 1 - 2 59 80
BC147324.1 - 2 - 3 184 80
BC147325.1 - 2 - 3 184 80
BQ939386.1 - 2 - 3 186 80

View Full Product Details