The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024920.1 | NP_001020091.1 | 3 - 4 | 228 | 118 |
GenBank mRNA | AK213971.1 | - | 2 - 3 | 101 | 118 |
BC147324.1 | - | 3 - 4 | 226 | 118 | |
BC147325.1 | - | 3 - 4 | 226 | 118 | |
BQ939386.1 | - | 3 - 4 | 228 | 118 |