The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027858.1 | 7 - 8 | 2337 | 76 | |
GenBank mRNA | BC141384.1 | - | 7 - 8 | 2333 | 76 |
DQ117602.1 | - | 7 - 8 | 2337 | 76 |