Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H13
Entrez Gene ID
14950
Gene Name
histocompatibility 13
Gene Aliases
1200006O09Rik, 4930443L17Rik, 5031424B04Rik, AV020344, H-13, Hm13, PSL3, Spp
Chromosome Location
Chr.2: 152669437 - 152708668 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159551.1 NP_001153023.1 10 - 11 1072 66
NM_010376.4 NP_034506.1 10 - 11 1072 66
GenBank mRNA AF483214.1 - 10 - 11 1039 66
AF515662.1 - 10 - 11 951 66
AJ345032.1 - 10 - 11 1082 66
AK007664.1 - 10 - 11 1054 66
AK012600.1 - 10 - 11 1065 66
AK028109.1 - 10 - 11 1041 66
AK049101.1 - 10 - 11 1046 66
AK050664.1 - 10 - 11 1015 66
AK077983.1 - 3 - 4 167 66
AK080966.1 - 10 - 11 1034 66
AK159176.1 - 10 - 11 1046 66
AK169922.1 - 10 - 11 1029 66
AK171790.1 - 9 - 10 875 66
BC034217.1 - 5 - 6 346 66
BC045195.1 - 6 - 7 428 66
DQ168449.1 - 10 - 11 1088 66

View Full Product Details