The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133202.2 | NP_573465.1 | 1 - 2 | 412 | 64 |
GenBank mRNA | AF283762.1 | - | 1 - 2 | 412 | 64 |
BC117988.2 | - | 1 - 2 | 391 | 64 | |
BC117989.2 | - | 1 - 2 | 391 | 64 |