Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tanc2
Entrez Gene ID
77097
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
Gene Aliases
3526402J09Rik, 5730590C14Rik
Chromosome Location
Chr.11: 105589986 - 105929304 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181071.3 NP_851416.2 12 - 13 2038 145
GenBank mRNA AK173111.1 - 3 - 4 508 145

View Full Product Details