The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252573.1 | NP_001239502.1 | 7 - 8 | 1001 | 74 |
NM_001252574.1 | NP_001239503.1 | 7 - 8 | 936 | 74 | |
NM_001252575.1 | NP_001239504.1 | 7 - 8 | 884 | 74 | |
NM_144893.2 | NP_659142.1 | 7 - 8 | 955 | 74 | |
NR_045544.1 | 8 - 9 | 832 | 74 | ||
GenBank mRNA | AK053180.1 | - | 7 - 8 | 871 | 74 |
AK155539.1 | - | 7 - 8 | 970 | 74 | |
AK160899.1 | - | 7 - 8 | 941 | 74 | |
AK170715.1 | - | 7 - 8 | 906 | 74 | |
BC018327.1 | - | 8 - 9 | 821 | 74 | |
BC094025.1 | - | 7 - 8 | 934 | 74 |