Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnttip1
Entrez Gene ID
76233
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
6430706C13Rik
Chromosome Location
Chr.2: 164745992 - 164768220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133763.1 NP_598524.1 9 - 10 700 97
GenBank mRNA AB045974.1 - 9 - 10 693 97
AK145239.1 - 9 - 10 726 97
AK168357.1 - 9 - 10 720 97
AK185487.1 - 3 - 4 127 97
BC003486.1 - 9 - 10 700 97

View Full Product Details