Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cep95
Entrez Gene ID
320162
Gene Name
centrosomal protein 95
Gene Aliases
4732496G21Rik, AI118346, AI448335, Ccdc45, D330027A14Rik, F630025I20Rik
Chromosome Location
Chr.11: 106789252 - 106818861 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166685.1 NP_001160157.1 14 - 15 1829 71
NM_177088.3 NP_796062.2 15 - 16 1958 71
GenBank mRNA AK076400.1 - 15 - 16 1936 71
BC090675.1 - 15 - 16 1914 71
BC116308.1 - 14 - 15 1795 71
BC116309.1 - 14 - 15 1812 71

View Full Product Details