The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037754.3 | NP_001032843.1 | 10 - 11 | 1257 | 73 |
NM_001037755.3 | NP_001032844.2 | 10 - 11 | 1377 | 73 | |
NM_130862.4 | NP_570932.2 | 10 - 11 | 1377 | 73 | |
GenBank mRNA | AF390178.1 | - | 10 - 11 | 1335 | 73 |
AF390179.1 | - | 10 - 11 | 1215 | 73 | |
AK049469.1 | - | 10 - 11 | 1378 | 73 | |
AK145924.1 | - | 10 - 11 | 1378 | 73 | |
AK158579.1 | - | 10 - 11 | 1399 | 73 | |
AK160401.1 | - | 10 - 11 | 1259 | 73 | |
AK170190.1 | - | 4 - 5 | 419 | 73 | |
BC006620.1 | - | 10 - 11 | 1368 | 73 | |
BC016411.1 | - | 10 - 11 | 1368 | 73 |