Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr3
Entrez Gene ID
234549
Gene Name
HEAT repeat containing 3
Gene Aliases
C030036P15Rik
Chromosome Location
Chr.8: 88137885 - 88171952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172757.3 NP_766345.3 13 - 14 1877 63
GenBank mRNA AK047782.1 - 13 - 14 1908 63
AK052635.1 - 13 - 14 1904 63
AK134510.1 - 13 - 14 1882 63
AK136424.1 - 13 - 14 1887 63
AK140522.1 - 13 - 14 1895 63
AK163715.1 - 13 - 14 2640 63
AK165541.1 - 13 - 14 1771 63
BC030373.1 - 9 - 10 1222 63
BC048074.1 - 2 - 3 595 63
BC065060.1 - 13 - 14 1877 63

View Full Product Details