Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfdc11
Entrez Gene ID
629761
Gene Name
WAP four-disulfide core domain 11
Gene Aliases
-
Chromosome Location
Chr.2: 164662894 - 164674253 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161806.1 NP_001155278.1 4 - 5 450 69
GenBank mRNA AK079019.1 - 4 - 5 451 69

View Full Product Details