The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033321.3 | NP_201578.2 | 1 - 2 | 206 | 77 |
GenBank mRNA | AF333331.1 | - | 1 - 2 | 212 | 77 |
AK134662.1 | - | 1 - 2 | 166 | 77 | |
BC138189.1 | - | 1 - 2 | 206 | 77 | |
BC145181.1 | - | 1 - 2 | 206 | 77 | |
JX570808.1 | - | 1 - 2 | 202 | 77 |