Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pecam1
Entrez Gene ID
18613
Gene Name
platelet/endothelial cell adhesion molecule 1
Gene Aliases
C85791, Cd31, PECAM-1, Pecam
Chromosome Location
Chr.11: 106654217 - 106750623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK037551.1 - 13 - 14 2634 116
AK084527.1 - 12 - 13 2224 116
BC008519.1 - 12 - 13 2228 116
BC085502.1 - 12 - 13 2030 116
L06039.1 - 12 - 13 2113 116
S79861.1 - 4 - 5 188 116

View Full Product Details