Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Card14
Entrez Gene ID
170720
Gene Name
caspase recruitment domain family, member 14
Gene Aliases
Bimp2, CARMA2
Chromosome Location
Chr.11: 119307096 - 119345375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130886.3 NP_570956.1 6 - 7 1168 71
GenBank mRNA AF363457.1 - 6 - 7 1147 71
AK028888.1 - 1 - 2 154 71
AK086176.1 - 7 - 8 1417 71
BC004692.1 - 3 - 4 603 71
BC029102.1 - 6 - 7 1168 71

View Full Product Details