Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gse1
Entrez Gene ID
382034
Gene Name
genetic suppressor element 1
Gene Aliases
2210013I18Rik, mKIAA0182
Chromosome Location
Chr.8: 120228826 - 120581383 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145896.1 NP_001139368.1 14 - 15 3551 118
NM_001145897.1 NP_001139369.1 14 - 15 3495 118
NM_198671.2 NP_941073.2 14 - 15 3683 118
GenBank mRNA AK033263.1 - 14 - 15 3498 118
AK042915.1 - 14 - 15 3500 118
AK122216.1 - 10 - 11 2725 118
AK136004.1 - 4 - 5 994 118
AK154829.1 - 14 - 15 3530 118
BC043094.1 - 14 - 15 3683 118

View Full Product Details