The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054095.2 | NP_473436.1 | 11 - 12 | 1386 | 76 |
GenBank mRNA | AF411253.1 | - | 11 - 12 | 1380 | 76 |
AK035168.1 | - | 9 - 10 | 759 | 76 | |
AK082727.1 | - | 11 - 12 | 996 | 76 | |
BC107384.1 | - | 12 - 13 | 981 | 76 | |
BC107385.1 | - | 12 - 13 | 945 | 76 | |
BC115857.1 | - | 12 - 13 | 918 | 76 | |
BC115963.1 | - | 12 - 13 | 906 | 76 |