The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144931.3 | NP_659180.1 | 5 - 6 | 397 | 100 |
GenBank mRNA | AK146202.1 | - | 5 - 6 | 355 | 100 |
AK162251.1 | - | 5 - 6 | 346 | 100 | |
AK166792.1 | - | 3 - 4 | 2923 | 100 | |
BC019163.1 | - | 5 - 6 | 377 | 100 | |
BC021510.1 | - | 5 - 6 | 378 | 100 | |
BC023897.1 | - | 5 - 6 | 354 | 100 | |
BC027124.1 | - | 5 - 6 | 341 | 100 |