The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027961.1 | NP_082237.1 | 3 - 4 | 553 | 70 |
GenBank mRNA | AK007299.1 | - | 3 - 4 | 418 | 70 |
BC116847.1 | - | 3 - 4 | 553 | 70 | |
BC116875.1 | - | 3 - 4 | 553 | 70 |