Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Casp1
Entrez Gene ID
12362
Gene Name
caspase 1
Gene Aliases
ICE, Il1bc
Chromosome Location
Chr.9: 5298517 - 5307281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009807.2 NP_033937.2 9 - 10 1411 83
GenBank mRNA AK132826.1 - 7 - 8 2123 83
BC008152.1 - 9 - 10 1411 83
L03799.1 - 8 - 9 1213 83
L28095.1 - 9 - 10 1257 83

View Full Product Details