The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164497.1 | NP_001157969.1 | 5 - 6 | 959 | 110 |
NM_001164498.1 | NP_001157970.1 | 5 - 6 | 933 | 110 | |
NM_001164499.1 | NP_001157971.1 | 5 - 6 | 959 | 110 | |
GenBank mRNA | AK077553.1 | - | 5 - 6 | 961 | 110 |
BC059053.1 | - | 5 - 6 | 932 | 110 | |
BC080314.1 | - | 4 - 5 | 534 | 110 | |
BC144796.1 | - | 5 - 6 | 917 | 110 | |
BC144797.1 | - | 5 - 6 | 917 | 110 | |
BC145737.1 | - | 5 - 6 | 917 | 110 |