Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpne2
Entrez Gene ID
234577
Gene Name
copine II
Gene Aliases
3322401K10Rik
Chromosome Location
Chr.8: 94533028 - 94570529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153507.2 NP_705727.1 8 - 9 983 101
GenBank mRNA AK160238.1 - 8 - 9 973 101
BC022128.1 - 8 - 9 944 101
BC023348.1 - 3 - 4 281 101
BC031801.1 - 8 - 9 983 101

View Full Product Details