The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024191.2 | NP_077153.1 | 5 - 6 | 658 | 118 |
NM_024269.4 | NP_077231.1 | 5 - 6 | 496 | 118 | |
GenBank mRNA | AK006418.1 | - | 5 - 6 | 483 | 118 |
AK006646.1 | - | 5 - 6 | 482 | 118 | |
AK018229.1 | - | 5 - 6 | 468 | 118 | |
AK149620.1 | - | 5 - 6 | 495 | 118 | |
AK150513.1 | - | 5 - 6 | 660 | 118 | |
AK155242.1 | - | 5 - 6 | 609 | 118 | |
AK161325.1 | - | 5 - 6 | 477 | 118 | |
BC002131.1 | - | 5 - 6 | 503 | 118 | |
BC024708.1 | - | 5 - 6 | 607 | 118 | |
CT010168.1 | - | 5 - 6 | 394 | 118 |