Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Katnb1
Entrez Gene ID
74187
Gene Name
katanin p80 (WD40-containing) subunit B 1
Gene Aliases
2410003J24Rik, KAT
Chromosome Location
Chr.8: 95081173 - 95099874 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028805.2 NP_083081.2 10 - 11 1221 74
GenBank mRNA AK010364.1 - 10 - 11 1152 74
AK031630.1 - 10 - 11 1133 74
AK034103.1 - 10 - 11 1141 74
AK037858.1 - - 1043 74
AK049345.1 - 10 - 11 1135 74
AK054173.1 - 10 - 11 1193 74
AK087973.1 - 10 - 11 1134 74
AK136322.1 - 10 - 11 1141 74
AK179072.1 - 1 - 2 118 74
BC045200.1 - 9 - 10 1132 74

View Full Product Details