Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog1
Entrez Gene ID
16834
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
6030445I15, Ldlb, mKIAA1381
Chromosome Location
Chr.11: 113649178 - 113669331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013581.3 NP_038609.3 6 - 7 1309 67
GenBank mRNA AF109377.2 - 6 - 7 1292 67
AK031510.1 - 5 - 6 765 67
AK034636.2 - 6 - 7 1314 67
AK152464.1 - 6 - 7 1300 67
AK159497.1 - 6 - 7 1311 67
AK162971.1 - 6 - 7 1310 67
AK171574.1 - 6 - 7 1303 67
AK191517.1 - 1 - 2 52 67
AK220387.1 - 6 - 7 1399 67
BC063056.1 - 6 - 7 1291 67

View Full Product Details