The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001491.1 | NP_001001491.1 | 4 - 5 | 482 | 113 |
GenBank mRNA | BC023827.1 | - | 4 - 5 | 476 | 113 |
BC032174.1 | - | 4 - 5 | 478 | 113 | |
BC032175.1 | - | 4 - 5 | 478 | 113 | |
BC070421.1 | - | 4 - 5 | 482 | 113 |