The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014395.2 | NP_001014417.1 | 6 - 7 | 816 | 98 |
GenBank mRNA | AK135808.1 | - | 4 - 5 | 533 | 98 |
AK162068.1 | - | 2 - 3 | 324 | 98 | |
BC084593.1 | - | 6 - 7 | 820 | 98 | |
CJ052642.1 | - | 4 - 5 | 533 | 98 |