Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wscd1
Entrez Gene ID
216881
Gene Name
WSC domain containing 1
Gene Aliases
mKIAA0523
Chromosome Location
Chr.11: 71749923 - 71789647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC030477.1 - 1 - 2 256 57

View Full Product Details