The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177618.4 | NP_808286.2 | 2 - 3 | 807 | 110 |
GenBank mRNA | AK220346.1 | - | 1 - 2 | 513 | 110 |
BC030477.1 | - | 2 - 3 | 945 | 110 | |
BC048831.1 | - | 1 - 2 | 626 | 110 | |
BC063070.1 | - | 2 - 3 | 807 | 110 |