Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aipl1
Entrez Gene ID
114230
Gene Name
aryl hydrocarbon receptor-interacting protein-like 1
Gene Aliases
A930007I01Rik
Chromosome Location
Chr.11: 72028717 - 72042375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053245.2 NP_444475.2 1 - 2 147 69
GenBank mRNA BC028285.1 - 1 - 2 147 69
CK618559.1 - 1 - 2 147 69

View Full Product Details