Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpain
Entrez Gene ID
69723
Gene Name
RPA interacting protein
Gene Aliases
2400006N03Rik, Rip
Chromosome Location
Chr.11: 70970200 - 70977933 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252413.1 NP_001239342.1 2 - 3 564 75
NM_001252414.1 NP_001239343.1 2 - 3 564 75
NM_001252415.1 NP_001239344.1 2 - 3 564 75
NM_027186.2 NP_081462.1 2 - 3 564 75
GenBank mRNA AK010289.1 - 2 - 3 293 75
BC047996.1 - 2 - 3 551 75
CB588111.1 - 2 - 3 564 75
DQ810267.1 - 2 - 3 257 75
DQ810268.1 - 2 - 3 257 75
DQ810269.1 - 2 - 3 257 75
DQ810270.1 - 2 - 3 257 75
DQ810271.1 - 2 - 3 257 75

View Full Product Details